Learn More
Aviva Systems Biology Corp CD6 Antibody - C-terminal region (ARP63343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD6. It was validated on Western Blot. . Alternate names: FLJ44171, TP120ApplicationWB. Species reactivity: Cow, Dog, Human, Rabbit, Rat
Catalog No. 50-287-944