Learn More
ACROBiosystems ACROBiosystems Human CD6 / TP120 Protein, His Tag
Catalog No. 501956415
Encompass_Preferred
Recombinant Protein;1MG;Human CD6, His Tag (CD6-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 18 - Glu 398 (Accession # P30203-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.5 kDa.
Catalog No. 50-195-6415 Supplier ACROBiosystems Supplier No. CD6H52H71MG
Recombinant Protein;1MG;Human CD6, His Tag (CD6-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 18 - Glu 398 (Accession # P30203-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.5 kDa.