Learn More
Genscript Corporation CD6 His Cynomolgus
Supplier:  Genscript Corporation Z04028500
Encompass
The cell surface glycoprotein CD6 is expressed on leukocytes and mediates T cell trafficking across endothelial cell barriers CD6 is a lymphocyte surface co-receptor physically associated with the T-cell receptor (TCR)/CD3 complex at the center of the immunological synapse
Catalog No. 50-000-58026