Learn More
Medchemexpress LLC CD6 Human HEK293 20ug
SDP
Catalog No. 5000182764
Encompass_Preferred
The CD6 protein is a cell adhesion molecule that mediates cell-cell contacts and regulates T cell responses through interaction with ALCAM/CD166 It functions as a costimulatory molecule promoting T cell activation proliferation and contributing to the formation of immune synapses CD6 Protein Human (HEK293 Fc) is the recombinant human-derived CD6 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-82764 Supplier Medchemexpress LLC Supplier No. HYP7292220UG
The CD6 protein is a cell adhesion molecule that mediates cell-cell contacts and regulates T cell responses through interaction with ALCAM/CD166 It functions as a costimulatory molecule promoting T cell activation proliferation and contributing to the formation of immune synapses CD6 Protein Human (HEK293 Fc) is the recombinant human-derived CD6 protein expressed by HEK293 with C-hFc labeled tag