Learn More
Biotium CD6 (Negative Marker of T-regulatory Cells)(SPV-L14), CF488A conjugate, 0.1mg/mL
Supplier:  Biotium BNC880402100
Encompass_Preferred
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular scavenger receptor cysteine-rich (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.
Catalog No. 50-252-3715