Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3690MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: Y2/51. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: U937, KG1a, HEL cells. Spleen. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Phytohemagglutinin (PHA)-stimulated peripheral blood cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 105kDa ;90kDa.Chromosome: 17q21.32. Gene Symbol: ITGB3. Gene ID: 3690. Ensemble: ENSG00000259207. Uniprot: NP_000203.
Catalog No. 50-188-4842