Learn More
Sino Biological CD63 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11271MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD63 (rh CD63; Catalog#11271-H08H; P08962-1; Ala 103-Val 203). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7405