Learn More
Santa Cruz Biotechnology Anti-CD63 Antibody (MX-49.129.5), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC5275
Encompass
Anti-CD63 Antibody (MX-49.129.5) is recommended for detection of CD63 of mouse, rat and human origin by WB, IP, IF, IHC(P), FCM and ELISA.
Catalog No. NC1297964