Learn More
FUJIFILM Biosciences CD63-Capture Human Exosome ELI
Catalog No. 5000450394
Encompass_Preferred
The CD63-Capture Human Exosome ELISA Kit Streptavidin HRP from FUJIFILM Wako is designed for the detection of CD63-positive human exosomes in various biological samples including serum plasma and culture supernatants This sandwich ELISA kit utilizes an anti-CD63 antibody-coated plate a biotinylated detection antibody anti-CD63 or user-provided and HRP-conjugated streptavidin for highly sensitive detection It does not require EV isolation and supports measurement of any vesicles presenting a surface marker for which a biotin-labeled antibody is available Ideal for researchers studying EV biology disease biomarkers or therapeutic vesicles CD9 CD81 ELISA are also available
Catalog No. 50-004-50394 Supplier FUJIFILM Biosciences Supplier No. 29083601
The CD63-Capture Human Exosome ELISA Kit Streptavidin HRP from FUJIFILM Wako is designed for the detection of CD63-positive human exosomes in various biological samples including serum plasma and culture supernatants This sandwich ELISA kit utilizes an anti-CD63 antibody-coated plate a biotinylated detection antibody anti-CD63 or user-provided and HRP-conjugated streptavidin for highly sensitive detection It does not require EV isolation and supports measurement of any vesicles presenting a surface marker for which a biotin-labeled antibody is available Ideal for researchers studying EV biology disease biomarkers or therapeutic vesicles CD9 CD81 ELISA are also available