Learn More
Medchemexpress LLC CD63 Human/Cynomolg 5ug
SDP
Catalog No. 5000182030
Encompass_Preferred
CD63 protein acts as a receptor for TIMP1 activating cell signaling pathways and promoting cell survival It plays a key role in integrin signaling leading to activation of AKT FAK/PTK2 and MAP kinases CD63 Protein Human/Cynomolgus (HEK293 N-His) is the recombinant human-derived CD63 protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-82030 Supplier Medchemexpress LLC Supplier No. HYP727155UG
CD63 protein acts as a receptor for TIMP1 activating cell signaling pathways and promoting cell survival It plays a key role in integrin signaling leading to activation of AKT FAK/PTK2 and MAP kinases CD63 Protein Human/Cynomolgus (HEK293 N-His) is the recombinant human-derived CD63 protein expressed by HEK293 with N-6 His labeled tag