Learn More
CAPRICO BIOTECHNOLOGIES INC CD64 Antibody, APC Conjugated
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4145045
Encompass_Preferred
The Caprico CD64 Antibody APC Conjugated specifically binds to human CD64 for flow cytometry and other assays. Available in 100, 500, or 1000 tests, it's optimized for staining blood cells but also works on other samples. Each vial has 100 μg of antibody in 0.2 ml buffer with sodium azide. Store at 2-8°C; stable for 12 months.
Catalog No. 50-227-9228