Learn More
Sino Biological Recombinant Human CD64/FCGR1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10256H08S1
Encompass_Preferred
A DNA sequence encoding the human FCGR1A (NP_000557.1) (Met1-Pro288) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5458