Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 968MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: LAMP4/824. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tonsil, Lymph Node or Spleen. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human LAMP4 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 110kDa.Chromosome: 17p13.1. Gene Symbol: CD68. Gene ID: 968. Ensemble: ENSG00000129226. Uniprot: NP_001035148.
Catalog No. 50-188-5374