Learn More
CAPRICO BIOTECHNOLOGIES INC CD68 FITC Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4064015
Encompass_Preferred
The CD68 FITC Antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes human CD68. It is conjugated to FITC and is suitable for flow cytometry, immunofluorescence, and immunohistochemistry. This antibody detects macrophages and monocytes. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The immunogen is human monocyte membranes. This antibody is recommended for detection of CD68 on human peripheral blood monocytes, macrophages, and dendritic cells by flow cytometry.
Catalog No. 50-227-9231