Learn More
Medchemexpress LLC CD68 Human sf9 Hi 5ug
SDP
Catalog No. 5000178943
Encompass_Preferred
CD68 protein is essential for phagocytic activities in tissue macrophages including lysosomal metabolism and interactions with cells and pathogens It binds to lectins or selectins aiding targeted migration of specific macrophage subsets CD68 s rapid recirculation from endosomes and lysosomes to the plasma membrane allows macrophages to crawl or interact with selectin-bearing substrates and other cells CD68 Protein Human (sf9 His) is the recombinant human-derived CD68 protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-78943 Supplier Medchemexpress LLC Supplier No. HYP742795UG
CD68 protein is essential for phagocytic activities in tissue macrophages including lysosomal metabolism and interactions with cells and pathogens It binds to lectins or selectins aiding targeted migration of specific macrophage subsets CD68 s rapid recirculation from endosomes and lysosomes to the plasma membrane allows macrophages to crawl or interact with selectin-bearing substrates and other cells CD68 Protein Human (sf9 His) is the recombinant human-derived CD68 protein expressed by Sf9 insect cells with C-His labeled tag