Learn More
Biocare Medical, LLC CD68 Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM033AA
Encompass_Preferred
The CD68 antibody antigen is a 110 kDa highly glycosylated transmembrane protein which is mainly located in lysosomes. CD68 is commonly regarded as a marker for monocytes and macrophages in many human tissues as well as fibroblasts, endothelial cells and tumor cells. Studies have shown that the CD68 antibody stains blast cells in a large percentage of acute myelogenous leukemia but none in acute lymphoblastic leukemia. Another study showed that [KP1] stained normal/reactive and neoplastic mast cells in lymph node and mastocytosis. The intensity of CD68 staining in individual cell types was found to depend on the fixation technique.
Catalog No. 50-830-11