Learn More
Medchemexpress LLC CD68 Rat HEK293 H 10ug
SDP
Catalog No. 5000178767
Encompass_Preferred
CD68 is a protein belonging to the LAMP (lysosome-associated membrane protein) family It is expressed primarily in macrophages monocytes and dendritic cells serving as a marker for these immune cells CD68 Protein Rat (HEK293 His) is the recombinant rat-derived CD68 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78767 Supplier Medchemexpress LLC Supplier No. HYP7427610UG
CD68 is a protein belonging to the LAMP (lysosome-associated membrane protein) family It is expressed primarily in macrophages monocytes and dendritic cells serving as a marker for these immune cells CD68 Protein Rat (HEK293 His) is the recombinant rat-derived CD68 protein expressed by HEK293 with C-His labeled tag