Learn More
Aviva Systems Biology Corp CD69 Antibody - middle region (ARP63035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD69. It was validated on Western Blot. . Alternate names: CLEC2C, AIM, EA1, MLR-3, GP32/28, BL-AC/P26ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-850