Learn More
Genscript Corporation CD69/CLEC2C His Human
Catalog No. 5000057021
Encompass
CLEC2C (CD69) is a membrane-bound type II C-lectin receptor and acts as a costimulatory molecule for T cell activation and proliferation It is involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes natural killer (NK) cells and platelets CLEC2C is a disulfide-linked homodimer protein with two differentially glycosylated subunits
Catalog No. 50-000-57021 Supplier Genscript Corporation Supplier No. Z04220100
CLEC2C (CD69) is a membrane-bound type II C-lectin receptor and acts as a costimulatory molecule for T cell activation and proliferation It is involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes natural killer (NK) cells and platelets CLEC2C is a disulfide-linked homodimer protein with two differentially glycosylated subunits