Learn More
CAPRICO BIOTECHNOLOGIES INC CD69 PE-Cyanine5 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4146075
Encompass_Preferred
The CD69 PE-Cyanine5 antibody is a mouse monoclonal antibody that binds to human CD69. It is conjugated to PE-Cyanine5 and is suitable for flow cytometry. This antibody recognizes an extracellular epitope of CD69 and can be used to identify activated lymphocytes. The antibody is supplied at 0.2 mg/mL in PBS with 0.1% sodium azide and 2 mg/mL BSA. The recommended dilution for flow cytometry is 5-25 μL per test. This antibody is guaranteed for 12 months from date of receipt when stored at 2-8°C.
Catalog No. 50-227-9233