Learn More
CAPRICO BIOTECHNOLOGIES INC CD69 PerCP-Cyanine5.5
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4146065
Encompass_Preferred
The CD69 PerCP-Cyanine5.5 antibody from Thermo Fisher Scientific is a mouse monoclonal antibody that recognizes human CD69. It is conjugated to PerCP-Cyanine5.5 and is recommended for flow cytometry analysis of human blood cells. This antibody detects the CD69 antigen, an early activation marker expressed on T and B lymphocytes, NK cells, granulocytes, and platelets. The CD69 PerCP-Cyanine5.5 antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100 μL of antibody. This product requires storage at 2-8°C and has a shelf life of 12 months from date of receipt.
Catalog No. 50-227-9232