Learn More
ACROBiosystems Recombinant Protein;Human CD69 / CLEC2C Protein;HEK293;100UG;CD9,CD69,CLEC2C,AIM,BL-AC/P26,EA1,GP32/28,MLR-3
Catalog No. 501658056
Encompass_Preferred
Recombinant Protein;Human CD69 / CLEC2C Protein;100UG;Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession # AAH07037).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8056 Supplier ACROBiosystems Supplier No. CD9H5222100UG
Recombinant Protein;Human CD69 / CLEC2C Protein;100UG;Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession # AAH07037).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 23 kDa and 26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.