Learn More
Biocare Medical, LLC CD7 Antibody - Concentrate, 0.5 ml
SDP
Supplier:  Biocare Medical, LLC CM158BK
Encompass_Preferred
The CD7 molecule is a membrane-bound glycoprotein of 40 kDa and is the earliest T-cell specific antigen to be expressed in lymphocytes. CD7 is expressed in the majority of thymocytes, peripheral blood T-cells and natural killer cells. Reports state that CD7 staining is significantly lower in mycosis fungoides than in benign dermatoses. Studies have shown that when used in combination with CD4, CD7 has been useful for differentiating mycosis fungoides or Sezary syndrome, both cutaneous T-cell lymphomas, from benign dermatoses.
Catalog No. 50-825-62