Learn More
Diagnostic Biosystems CD7 CD7 (CBC.37.80)
Supplier:  Diagnostic Biosystems PDM599
Encompass_Preferred
CD7 is expressed by most peripheral blood T cells, NK cells, and all thymocytes. It is one of the earliest surface antigens on T and NK-cell lineages. The antibody is a useful aid for classification of T-cell malignancies.
Catalog No. 50-240-7574