Learn More
CAPRICO BIOTECHNOLOGIES INC CD7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4025175
Encompass_Preferred
The CD7 Antibody is a mouse monoclonal antibody that recognizes human CD7. CD7 is a 40-60 kDa transmembrane glycoprotein found on thymocytes and mature T cells. This antibody is unconjugated and is recommended for detection of CD7 protein in Western blotting, immunoprecipitation, flow cytometry, and immunohistochemistry. The antibody is supplied as 100 μg purified antibody in PBS with 0.09% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9241