Learn More
Sino Biological Recombinant Anti-CD7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50393R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD7 / LEU9 extracellular domain (rM CD7; Catalog#50393-M08H; NP_033984.1; Met 1-Pro 150).
Catalog No. 50-224-8396