Learn More
Sino Biological Anti-CD7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50393RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD7 / LEU9 extracellular domain (rM CD7; Catalog#50393-M08H; NP_033984.1; Met 1-Pro 150). CD7 specific IgG was purified by mouse CD7 affinity chromatography.
Catalog No. 50-224-8109