Learn More
ACROBiosystems ACROBiosystems Human CD7 Protein, Llama IgG2b Fc Tag, low endotoxin
Catalog No. 501956609
Encompass_Preferred
Recombinant Protein;100UG;Human CD7, Llama IgG2b Fc Tag, low endotoxin (CD7-H5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a llama IgG2b Fc tag at the N-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD7, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD7-H5258) at 5 μg/mL (100 μL/well) can bind Human SECTM1, His Tag (Cat. No. SE1-H5227) with a linear range of 10-39 ng/mL (QC tested).
Catalog No. 50-195-6609 Supplier ACROBiosystems Supplier No. CD7H5258100UG
Recombinant Protein;100UG;Human CD7, Llama IgG2b Fc Tag, low endotoxin (CD7-H5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a llama IgG2b Fc tag at the N-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD7, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD7-H5258) at 5 μg/mL (100 μL/well) can bind Human SECTM1, His Tag (Cat. No. SE1-H5227) with a linear range of 10-39 ng/mL (QC tested).