Learn More
CAPRICO BIOTECHNOLOGIES INC CD7 MF450, 4025145
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4025145
Encompass_Preferred
The CD7-MF450-4025145 is a monoclonal antibody from Caprico Biotechnologies. It is an IgG1 isotype antibody produced in mouse and has unknown cross-reactivity. The antibody is specific for human CD7, also known as gp40, and is recommended for use in flow cytometry, immunohistochemistry, and Western blot. The immunogen is human CD7 extracellular domain fusion protein. This antibody is supplied at 0.5 mg/ml in PBS with 0.02% sodium azide and should be stored at 4°C.
Catalog No. 50-227-9240