Learn More
Medchemexpress LLC CD7 Rat HEK293 Fc 10ug
SDP
Supplier:  Medchemexpress LLC HYP7427310UG
Encompass_Preferred
The CD7 Protein is a member of the immunoglobulin superfamily expressed in thymocytes and mature T cells CD7 Protein can activate the PI3K signaling pathway involved in the activation of T and NK cells CD7 Protein is a key factor in the treatment of T-cell acute lymphoblastic leukemia (T-ALL) and T-lymphoma CD7 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD7 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-78948