Learn More
Aviva Systems Biology Corp CD70 Antibody - N-terminal region (ARP61309_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61309_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD70. It was validated on Western Blot. . Alternate names: CD27L, CD27LG, TNFSF7ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-287-338