Learn More
Sino Biological Human CD70 / CD27L / TNFSF7 Protein (Fc & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological 10780H42HB100
Encompass_Preferred
A DNA sequence encoding the human CD70 (NP_001243.1) (Gln39-Pro193) was expressed with a N-terminal Fc region of human IgG1 tagged AVI tag at the N-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5234