Learn More
Medchemexpress LLC CD70 Rat HEK293 F 50ug
SDP
Catalog No. 5000182722
Encompass_Preferred
CD70 is the ligand of CD27 in activated T and B lymphocytes is an important cytokine in CD70-CD27 pathway involving with the generation and maintenance of T cell immunity[1] CD70 is a surface antigen also shows antiviral activity and regulates viability of tumor cells and regulatory T cells[2][3] As for CD70 protein in rat contains TNF 2 domain (57-188 a a) and transmembrane helix belonging to the tumor necrosis factor (TNF) family CD70 Protein Rat (HEK293 Fc) is 150 amino acids in length (Q46-P195) and is expressed in the HEK293 cells with N terminal hFc-tag
Catalog No. 50-001-82722 Supplier Medchemexpress LLC Supplier No. HYP7292950UG
CD70 is the ligand of CD27 in activated T and B lymphocytes is an important cytokine in CD70-CD27 pathway involving with the generation and maintenance of T cell immunity[1] CD70 is a surface antigen also shows antiviral activity and regulates viability of tumor cells and regulatory T cells[2][3] As for CD70 protein in rat contains TNF 2 domain (57-188 a a) and transmembrane helix belonging to the tumor necrosis factor (TNF) family CD70 Protein Rat (HEK293 Fc) is 150 amino acids in length (Q46-P195) and is expressed in the HEK293 cells with N terminal hFc-tag