Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7037MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: TFRC/1817. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: PC3 cells. Human placenta or breast carcinoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human TFRC protein fragment (aa94-212). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 85-95kDa (monomer) ;190kDa (dimer).Chromosome: 3q29. Gene Symbol: TFRC. Gene ID: 7037. Ensemble: ENSG00000072274. Uniprot: NP_001300894.
Catalog No. 50-188-5201