Learn More
Biolegend PE/Cyanine7 anti-human CD71
Supplier:  Biolegend 334112
Encompass_Preferred
PE/Cyanine7 anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC0672188