Learn More
ACROBiosystems ACROBiosystems Mouse CD73 / NT5E Protein, His Tag
Catalog No. 501957017
Encompass_Preferred
Recombinant Protein;100UG;Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 29 - Ser 551 (Accession # Q61503-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.9 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7017 Supplier ACROBiosystems Supplier No. CD3M52H9100UG
Recombinant Protein;100UG;Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 29 - Ser 551 (Accession # Q61503-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.9 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.