Learn More
Medchemexpress LLC HY-103695 1mg Medchemexpress, CD73-IN-1 CAS:2132396-40-6 Purity:>98%
SDP
Catalog No. 501968163
Encompass_Preferred
Medchemexpress, HY-103695 1mg CD73-IN-1 CAS:2132396-40-6 CD73-IN-1 is an inhibitor of CD73 which can be used in the treatment of cancer extracted from patent WO 2017153952 A1, example 80. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8163 Supplier Medchemexpress LLC Supplier No. HY1036951MG
Medchemexpress, HY-103695 1mg CD73-IN-1 CAS:2132396-40-6 CD73-IN-1 is an inhibitor of CD73 which can be used in the treatment of cancer extracted from patent WO 2017153952 A1, example 80. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.