Learn More
Medchemexpress LLC CD73-IN-3 | 2375815-63-5 | 99.9% | 286.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137246100MG
Encompass_Preferred
CD73-IN-3 is a potent inhibitor of CD73, demonstrating an IC50 of 7.3 nM in a Calu6 human cell assay. This compound, identified as example 2 from patent WO2019168744 A1, holds potential for cancer research. It inhibits CD73 activity in human serum with an EC50 of 0.213 μM. The product is a solid, white to off-white in color.
  • Potent inhibitor of CD73
  • IC50 of 7.3 nM in Calu6 human cell assay
  • Inhibits CD73 activity in human serum with an EC50 of 0.213 μM
  • Potential for cancer research
  • Appears as a solid, white to off-white
Catalog No. 50-003-48774