Learn More
Enzo Life Sciences CD73 monoclonal antibody (AT89E9) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7190100
Encompass_Preferred
Clone: AT89E9. Isotype: IgG2bκ. Immunogen: Recombinant human CD73 (aa 27-252) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3260