Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 972MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: LN-2. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Daudi or Raji Cells. Tonsil or lymph node. Cellular Staining: Cell surface, paranuclear globular. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: SU-DHL-4 lymphoma cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 33-41kDa.Chromosome: 5q33.1. Gene Symbol: CD74. Gene ID: 972. Ensemble: ENSG00000019582. Uniprot: XP_016865578.
Catalog No. 50-188-5376