Learn More
Genscript Corporation CD74/DHLAG His Mouse
Catalog No. 5000056937
Encompass
CD74 (invariant MHC class II) regulates protein trafficking and is a receptor for macrophage migration inhibitory factor (MIF) and d-dopachrome tautomerase (d-DT/MIF-2) CD74 expression is increased in tubular cells and/or glomerular podocytes and parietal cells in human metabolic nephropathies polycystic kidney disease graft rejection and kidney cancer and in experimental diabetic nephropathy and glomerulonephritis
Catalog No. 50-000-56937 Supplier Genscript Corporation Supplier No. Z04328500
CD74 (invariant MHC class II) regulates protein trafficking and is a receptor for macrophage migration inhibitory factor (MIF) and d-dopachrome tautomerase (d-DT/MIF-2) CD74 expression is increased in tubular cells and/or glomerular podocytes and parietal cells in human metabolic nephropathies polycystic kidney disease graft rejection and kidney cancer and in experimental diabetic nephropathy and glomerulonephritis