Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD74 Antibody FITC
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4088015
Encompass_Preferred
The Caprico Anti-CD74 Antibody FITC is a monoclonal antibody that specifically binds to CD74, also known as the invariant chain of MHC class II. It is conjugated to FITC, a fluorescent dye, allowing detection by flow cytometry. This antibody can be used to identify cells expressing CD74, including B cells, monocytes, macrophages, dendritic cells, and Langerhans cells. It is suitable for immunocytochemistry, immunohistochemistry, and flow cytometry applications. The antibody is sourced from mouse and supplied at 0.5 mg/mL in PBS with 0.1% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9249