Learn More
ACROBiosystems Recombinant Protein;Human CD74 / DHLAG Protein;HEK293;100UG;CD4,CD74,DHLAG,HLADG,II,Ia-GAMMA,p33,CLIP,INVG34
Supplier:  ACROBiosystems CD4H524C100UG
Encompass_Preferred
Recombinant Protein;Human CD74 / DHLAG Protein;100UG;Human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession # NP_004346.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 23-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7967