Learn More
Sino Biological Recombinant Human CD79 Protein (Flag & His Tag), HPLC-verified
Supplier:  Sino Biological CT081H2508H100
Encompass_Preferred
A DNA sequence encoding the human CD79A (NP_001774.1) (Met1-Arg143) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding human CD79B (NP_000617.1) (Met1-Asp159) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD79A & CD79B heterodimer was purified.
Catalog No. 50-211-4763