Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 973MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: IGA/764. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human IGA protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 44kDa.Chromosome: 19q13.2. Gene Symbol: CD79A. Gene ID: 973. Ensemble: ENSG00000105369. Uniprot: NP_067612.
Catalog No. 50-188-5384