Learn More
Aviva Systems Biology Corp CD79A Antibody - N-terminal region (ARP63188_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD79A. It was validated on Western Blot. . Alternate names: IGA, MB-1ApplicationWB. Species reactivity: Human
Catalog No. 50-287-906