Learn More
CAPRICO BIOTECHNOLOGIES INC CD79a Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4151045
Encompass_Preferred
The CD79a Antibody is a mouse monoclonal IgG1 antibody that recognizes human CD79a. CD79a is a component of the B cell antigen receptor complex and is expressed on the surface of B cells. This antibody is unconjugated and can be used for applications like flow cytometry, immunohistochemistry, and Western blot. The antibody is supplied as 100 μg purified antibody in PBS with 0.09% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9251