Learn More
Enzo Life Sciences CD79a Recombinant monoclonal antibody (IGA/1688R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3290100
Encompass_Preferred
Clone: IGA/1688R. Isotype: IgGκ. Immunogen: Synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a. Species reactivity: Human, Mouse, Rat. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2860