Learn More
Genscript Corporation CD79B His Mouse
Catalog No. 5000057682
Encompass
CD79B (also known as B29 Ig beta and B cell antigen receptor complex-associated protein beta-chain) is a 36-40 kDa member of the Ig-Superfamily It is required in cooperation with CD79A for initiation of the signal transduction cascade activated by the B-cell antigen receptor complex (BCR) which leads to internalization of the complex trafficking to late endosomes and antigen presentation Enhances phosphorylation of CD79A possibly by recruiting kinases which phosphorylate CD79A or by recruiting proteins which bind to CD79A and protect it from dephosphorylation
Catalog No. 50-000-57682 Supplier Genscript Corporation Supplier No. Z04329500
CD79B (also known as B29 Ig beta and B cell antigen receptor complex-associated protein beta-chain) is a 36-40 kDa member of the Ig-Superfamily It is required in cooperation with CD79A for initiation of the signal transduction cascade activated by the B-cell antigen receptor complex (BCR) which leads to internalization of the complex trafficking to late endosomes and antigen presentation Enhances phosphorylation of CD79A possibly by recruiting kinases which phosphorylate CD79A or by recruiting proteins which bind to CD79A and protect it from dephosphorylation