Learn More
Medchemexpress LLC CD79B Human Biotin 100ug
SDP
Catalog No. 5000196979
Encompass_Preferred
Recombinant human CD79B (extracellular residues Ala29-Asp159) produced in HEK293 cells and enzymatically biotinylated via an Avi tag. The construct includes a C-terminal His-Avi fusion, is >95% pure, and migrates ~35-45 kDa on SDS-PAGE due to glycosylation. Supplied with storage and reconstitution guidance for use in immobilization and binding assays.
  • Site-specific biotinylation for consistent immobilization.
  • C-terminal His-Avi tag for purification and controlled biotinylation.
  • Mammalian expression preserves native glycosylation and folding.
  • High purity suitable for ELISA, SPR/BLI, and flow cytometry.
  • Storage and reconstitution recommendations to maintain activity.
Catalog No. 50-001-96979 Supplier Medchemexpress LLC Supplier No. HYP70768100UG
Recombinant human CD79B (extracellular residues Ala29-Asp159) produced in HEK293 cells and enzymatically biotinylated via an Avi tag. The construct includes a C-terminal His-Avi fusion, is >95% pure, and migrates ~35-45 kDa on SDS-PAGE due to glycosylation. Supplied with storage and reconstitution guidance for use in immobilization and binding assays.
  • Site-specific biotinylation for consistent immobilization.
  • C-terminal His-Avi tag for purification and controlled biotinylation.
  • Mammalian expression preserves native glycosylation and folding.
  • High purity suitable for ELISA, SPR/BLI, and flow cytometry.
  • Storage and reconstitution recommendations to maintain activity.